Assay Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655004.1 | XP_007653194.1 | 3 - 4 | 798 | 61 | |
GenBank mRNA | JI879388.1 | - | 2 - 3 | 249 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
