Assay Details
Gene Symbol: Tmem180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648683.1 | XP_007646873.1 | 3 - 4 | 444 | 87 | |
XM_007648684.1 | XP_007646874.1 | 3 - 4 | 444 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
