Assay Details
| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503923.2 | XP_003503971.1 | 2 - 3 | 627 | 73 | |
| XM_007644456.1 | XP_007642646.1 | 2 - 3 | 353 | 73 | ||
| XM_007644457.1 | XP_007642647.1 | 2 - 3 | 414 | 73 | ||
More Information
Set Membership: |
|

Aqp1
Genomic Map