Assay Details
| Gene Symbol: Mrpl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514533.2 | XP_003514581.1 | 3 - 4 | 865 | 70 | |
| GenBank mRNA | JI893808.1 | - | 2 - 3 | 653 | 70 | |
More Information
Set Membership: |
|

Mrpl44
Genomic Map