Assay Details
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007638529.1 | XP_007636719.1 | 2 - 3 | 333 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
