Assay Details
| Gene Symbol: Trmt61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651511.1 | XP_007649701.1 | 2 - 3 | 359 | 63 | |
| GenBank mRNA | JP051827.1 | - | 1 - 2 | 112 | 63 | |
More Information
Set Membership: |
|

Trmt61b
Genomic Map