Assay Details
| Gene Symbol: LOC100752705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007639527.1 | XP_007637717.1 | 25 - 26 | 3554 | 106 | |
| XM_007639536.1 | XP_007637726.1 | 24 - 25 | 3353 | 106 | ||
More Information
Set Membership: |
|

LOC100752705
Genomic Map