Assay Details
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643039.1 | XP_007641229.1 | 3 - 4 | 282 | 74 | |
GenBank mRNA | JI890390.1 | - | 3 - 4 | 427 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
