Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 1 - 2 | 97 | 65 | |
| GenBank mRNA | JI883467.1 | - | 1 - 2 | 95 | 65 | |
| JP055927.1 | - | 1 - 2 | 95 | 65 | ||
More Information
Set Membership: |
|

LOC100758930
Genomic Map