Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 2 - 3 | 849 | 109 | |
| XM_007638093.1 | XP_007636283.1 | 2 - 3 | 996 | 109 | ||
| XM_007638097.1 | XP_007636287.1 | 2 - 3 | 858 | 109 | ||
| GenBank mRNA | JI883467.1 | - | 2 - 3 | 847 | 109 | |
| JP055927.1 | - | 2 - 3 | 847 | 109 | ||
More Information
Set Membership: |
|

LOC100758930
Genomic Map