Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 3 - 4 | 1118 | 79 | |
| XM_007638093.1 | XP_007636283.1 | 3 - 4 | 1265 | 79 | ||
| XM_007638097.1 | XP_007636287.1 | 3 - 4 | 1127 | 79 | ||

LOC100758930
Genomic Map