Assay Details
| Gene Symbol: LOC100758930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499079.2 | XP_003499127.1 | 7 - 8 | 1932 | 74 | |
| XM_007638093.1 | XP_007636283.1 | 7 - 8 | 2079 | 74 | ||
| XM_007638097.1 | XP_007636287.1 | 7 - 8 | 1941 | 74 | ||
| GenBank mRNA | JI862146.1 | - | 3 - 4 | 478 | 74 | |
| JP060170.1 | - | 3 - 4 | 478 | 74 | ||

LOC100758930
Genomic Map