Assay Details
| Gene Symbol: Rps27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503975.2 | XP_003504023.1 | 5 - 6 | 403 | 66 | |
| GenBank mRNA | JI884695.1 | - | 4 - 5 | 440 | 66 | |
More Information
Set Membership: |
|

Rps27a
Genomic Map