Assay Details
| Gene Symbol: Ppp1r15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505757.2 | XP_003505805.1 | 1 - 2 | 1760 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r15b| Gene Symbol: Ppp1r15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505757.2 | XP_003505805.1 | 1 - 2 | 1760 | 91 | |
Set Membership: |
|