Assay Details
Gene Symbol: Cbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647240.1 | XP_007645430.1 | 2 - 3 | 331 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647240.1 | XP_007645430.1 | 2 - 3 | 331 | 59 |
Set Membership: |
![]() ![]() ![]() ![]() |