Assay Details
| Gene Symbol: LOC100750934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655046.1 | XP_007653236.1 | 3 - 4 | 810 | 74 | |
| GenBank mRNA | JI891896.1 | - | 2 - 3 | 496 | 74 | |
More Information
Set Membership: |
|

LOC100750934
Genomic Map