Assay Details
Gene Symbol: Rbmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640570.1 | XP_007638760.1 | 2 - 3 | 194 | 75 | |
GenBank mRNA | JI896673.1 | - | 2 - 3 | 312 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
