Assay Details
Gene Symbol: Cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498306.2 | XP_003498354.1 | 6 - 7 | 2000 | 60 | |
GenBank mRNA | JI864968.1 | - | 6 - 7 | 1483 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
