Assay Details
| Gene Symbol: Ccdc28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641401.1 | XP_007639591.1 | 2 - 3 | 702 | 66 | |
| XM_007641402.1 | XP_007639592.1 | 2 - 3 | 702 | 66 | ||
| XM_007641403.1 | XP_007639593.1 | 3 - 4 | 836 | 66 | ||
More Information
Set Membership: |
|

Ccdc28b
Genomic Map