Assay Details
| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497373.2 | XP_003497421.1 | 1 - 2 | 133 | 63 | |
| XM_007611294.1 | XP_007609484.1 | 1 - 2 | 139 | 63 | ||
| GenBank mRNA | JI894936.1 | - | 1 - 2 | 261 | 63 | |
| JP039249.1 | - | 1 - 2 | 261 | 63 | ||
More Information
Set Membership: |
|

Mmachc
Genomic Map