Assay Details
Gene Symbol: Mrps34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501492.2 | XP_003501540.2 | 2 - 3 | 455 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
