Assay Details
Gene Symbol: Tmed2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648402.1 | XP_007646592.1 | 2 - 3 | 462 | 64 | |
GenBank mRNA | JI873289.1 | - | 2 - 3 | 457 | 64 | |
JP059525.1 | - | 2 - 3 | 457 | 64 | ||
U26264.1 | - | 462 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
