Assay Details
Gene Symbol: LOC100760475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641529.1 | XP_007639719.1 | 12 - 13 | 1476 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
