Assay Details
| Gene Symbol: Rps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497386.2 | XP_003497434.1 | 5 - 6 | 626 | 68 | |
| GenBank mRNA | JI871269.1 | - | 5 - 6 | 594 | 68 | |
| JP065117.1 | - | 5 - 6 | 594 | 68 | ||
More Information
Set Membership: |
|

Rps8
Genomic Map