Assay Details
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499078.2 | XP_003499126.2 | 6 - 7 | 677 | 64 | |
GenBank mRNA | JI877045.1 | - | 4 - 5 | 371 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
