Assay Details
Gene Symbol: Hn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642176.1 | XP_007640366.1 | 2 - 3 | 363 | 58 | |
GenBank mRNA | JP043540.1 | - | 2 - 3 | 363 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
