Assay Details
| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007623236.1 | XP_007621426.1 | 4 - 5 | 585 | 60 | |
| XM_007623241.1 | XP_007621431.1 | 4 - 5 | 584 | 60 | ||
| XM_007623249.1 | XP_007621439.1 | 4 - 5 | 589 | 60 | ||
| GenBank mRNA | JP044993.1 | - | 3 - 4 | 246 | 60 | |
More Information
Set Membership: |
|

Grk6
Genomic Map