Assay Details
Gene Symbol: Igll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643451.1 | XP_007641641.1 | 1 - 2 | 179 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
