Assay Details
| Gene Symbol: LOC100759441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647019.1 | XP_007645209.1 | 11 - 12 | 1711 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100759441| Gene Symbol: LOC100759441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647019.1 | XP_007645209.1 | 11 - 12 | 1711 | 90 | |
Set Membership: |
|