Assay Details
Gene Symbol: Icam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642520.1 | XP_007640710.1 | 2 - 3 | 458 | 65 | |
XM_007642521.1 | XP_007640711.1 | 3 - 4 | 393 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
