Assay Details
Gene Symbol: Pla2g12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503748.1 | XP_003503796.1 | 3 - 4 | 467 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
