Assay Details
Gene Symbol: LOC100751499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512386.2 | XP_003512434.1 | 3 - 4 | 460 | 99 | |
GenBank mRNA | JI862118.1 | - | 1 - 2 | 129 | 99 | |
JP048190.1 | - | 1 - 2 | 129 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
