Assay Details
| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651267.1 | XP_007649457.1 | 2 - 3 | 245 | 115 | |
| GenBank mRNA | JI868776.1 | - | 2 - 3 | 245 | 115 | |
More Information
Set Membership: |
|

Aunip
Genomic Map