Assay Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496246.2 | XP_003496294.1 | 4 - 5 | 636 | 79 | |
| GenBank mRNA | JI875619.1 | - | 3 - 4 | 528 | 79 | |
| JP053765.1 | - | 3 - 4 | 528 | 79 | ||
More Information
Set Membership: |
|

Rala
Genomic Map