Assay Details
Gene Symbol: Agbl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496515.1 | XP_003496563.1 | 8 - 9 | 849 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
