Assay Details
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508513.1 | XP_003508561.1 | 3 - 4 | 801 | 75 | |
XM_007649119.1 | XP_007647309.1 | 3 - 4 | 801 | 75 | ||
XM_007649120.1 | XP_007647310.1 | 3 - 4 | 801 | 75 | ||
XM_007649121.1 | XP_007647311.1 | 4 - 5 | 1016 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
