Assay Details
Gene Symbol: Sfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508298.2 | XP_003508346.1 | 1 | 823 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
