Assay Details
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644333.1 | XP_007642523.1 | 3 - 4 | 397 | 68 | |
GenBank mRNA | JI886536.1 | - | 3 - 4 | 363 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
