Assay Details
Gene Symbol: Cav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641504.1 | XP_007639694.1 | 1 - 2 | 337 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
