Assay Details
Gene Symbol: Ca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499339.2 | XP_003499387.1 | 2 - 3 | 109 | 69 | |
XM_007640106.1 | XP_007638296.1 | 2 - 3 | 109 | 69 | ||
XM_007640108.1 | XP_007638298.1 | 2 - 3 | 109 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
