Assay Details
Gene Symbol: LOC100763162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607746.1 | XP_007605936.1 | 2 - 3 | 325 | 80 | |
GenBank mRNA | JP041678.1 | - | 2 - 3 | 325 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
