Assay Details
| Gene Symbol: Rpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653071.1 | XP_007651261.1 | 404 | 77 | ||
| GenBank mRNA | JI884690.1 | - | 1 | 137 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl22| Gene Symbol: Rpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653071.1 | XP_007651261.1 | 404 | 77 | ||
| GenBank mRNA | JI884690.1 | - | 1 | 137 | 77 | |
Set Membership: |
|