Assay Details
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498305.2 | XP_003498353.1 | 4 - 5 | 719 | 51 | |
GenBank mRNA | JI877934.1 | - | 1 - 2 | 53 | 51 | |
JP055362.1 | - | 1 - 2 | 53 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() |
