Assay Details
| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509081.2 | XP_003509129.1 | 1 - 2 | 238 | 63 | |
| GenBank mRNA | JI886479.1 | - | 1 - 2 | 223 | 63 | |
| JP059029.1 | - | 1 - 2 | 223 | 63 | ||
More Information
Set Membership: |
|

Ndufb5
Genomic Map