Assay Details
| Gene Symbol: Atp5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654507.1 | XP_007652697.1 | 2 - 3 | 325 | 94 | |
| GenBank mRNA | JI891490.1 | - | 2 - 3 | 325 | 94 | |
More Information
Set Membership: |
|

Atp5f1
Genomic Map