Assay Details
| Gene Symbol: Papd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642344.1 | XP_007640534.1 | 7 - 8 | 889 | 65 | |
| XM_007642345.1 | XP_007640535.1 | 7 - 8 | 889 | 65 | ||
| GenBank mRNA | JI884938.1 | - | 7 - 8 | 881 | 65 | |
More Information
Set Membership: |
|

Papd7
Genomic Map