Assay Details
| Gene Symbol: Toe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497377.2 | XP_003497425.1 | 5 - 6 | 600 | 69 | |
| GenBank mRNA | JI895068.1 | - | 5 - 6 | 523 | 69 | |
| JP056733.1 | - | 5 - 6 | 523 | 69 | ||
More Information
Set Membership: |
|

Toe1
Genomic Map