Assay Details
Gene Symbol: Fgf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498395.2 | XP_003498443.1 | 1 - 2 | 350 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
