Assay Details
| Gene Symbol: Qdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505979.2 | XP_003506027.1 | 5 - 6 | 647 | 58 | |
| GenBank mRNA | JI887415.1 | - | 5 - 6 | 626 | 58 | |
| JP047660.1 | - | 5 - 6 | 626 | 58 | ||
More Information
Set Membership: |
|

Qdpr
Genomic Map