Assay Details
Gene Symbol: LOC100755875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647165.1 | XP_007645355.1 | 16 - 17 | 1685 | 66 | |
GenBank mRNA | JP038131.1 | - | 16 - 17 | 1685 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
